missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Leptin Protein
Recombinant Protein
£80.80 - £990.00
Specifications
Accession Number | NP_032519.1 |
---|---|
Gene ID (Entrez) | 16846 |
Name | Leptin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15956109
|
enQuireBio™
QP5429-200UG |
200 μg |
£80.80
200µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936109
|
enQuireBio™
QP5429-1000UG |
1 mg |
£191.00
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966109
|
enQuireBio™
QP5429-5MG |
5 mg |
£550.00
5mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946109
|
enQuireBio™
QP5429-10MG |
10 mg |
£990.00
10mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_032519.1 | |
Leptin Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MVPIQKVQDD TKTLIKTIVT RINDISHTQS VSAKQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQVLTSLP SQNVLQIAND LENLRDLLHL LAFSKSCSLP QTSGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDVSPEC | |
Greater than 95% as determined by SDS-PAGE. |
16846 | |
Research Use Only | |
LEP | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |