missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LBP Protein
Click to view available options
Quantity:
10 μg
2 μg
5 μg
Unit Size:
10µg
2µg
5µg
Specifications
Specifications
| Gene ID (Entrez) | 16803 |
| Name | LBP Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Gene Symbol | Lbp |
| Biological Activity | Up to 2microg/ml LBP mediates binding of FITC-LPS (0.5microg/ml) to CD14+CHO transfectants (2 x 106/ml). |
| Product Type | Recombinant Protein |
| Cross Reactivity | Mouse |
| Species | Mouse |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction