missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse LBP Protein

Product Code. 15985028
Click to view available options
Quantity:
10 μg
2 μg
5 μg
Unit Size:
10µg
2µg
5µg
This item is not returnable. View return policy

Product Code. 15985028

Brand: enQuireBio™ QP1074210ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the LBP was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 16803
Name LBP Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol Lbp
Biological Activity Up to 2microg/ml LBP mediates binding of FITC-LPS (0.5microg/ml) to CD14+CHO transfectants (2 x 106/ml).
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Mouse
Protein Tag His
Sequence MKAGTGPLLS TLLGLLFLSI QGTGGVNPGV VARITDKGLA YAAKEGLVAL QRELYKITLP DFSGDFKIKA VGRGQYEFHS LEIQNCELRG SSLKLLPGQG LSLAISDSSI GVRGKWKVRK SFLKLHGSFD LDVKGVTISV DLLLGMDPSG RPTVSASGCS SRICDLDVHI SGNVGWLLNL FHNQIESKLQ KVLENKVCEM IQKSVTSDLQ PYLQTLPVTA EIDNVLGIDY SLVAAPQAKA QVLDVMFKGE IFNRNHRSPV ATPTPTMSLP EDSKQMVYFA ISDYAFNIAS RVYHQAGYLN FSITDDMLPH DSGIRLNTKA FRPFTPQIYK KYPDMKLELL GTVVSAPILN VSPGNLSLAP QMEIEGFVIL PTSAREPVFR LGVVTNVFAS LTFNNSKVTG MLHPDKAQVR LIESKVGMFN VNLFQAFLNY YLLNSLYPDV NAELAQGFPL PLPRHIQLHD LDFQIRKDFL YLGANVQYMR V
Buffer Recombinant Mouse LBP was lyophilized from a protein solution (1 mg/ml) containing phosphate-buffered saline, pH 7.2.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.