missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse KGF/FGF-7 Protein

Product Code. 18446511 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
500 μg
Unit Size:
1000µg
100µg
10µg
500µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18446511 500 μg 500µg
18408381 10 μg 10µg
18475282 100 μg 100µg
18418381 1000 μg 1000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18446511 Supplier Novus Biologicals™ Supplier No. NBP235149500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

FGF7 Protein is fully biologically active when compared to standard. The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 10 ng/mL, corresponding to a specific activity of >1.0 x 10^5 IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P36363
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered solution in 20 mM PB, pH 8.0, 1 M NaCl.
Gene ID (Entrez) 2252
Molecular Weight (g/mol) M.W. (Theoretical): 18.7 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/μg of KGF/FGF-7 as determined by LAL method.
Gene Alias FGF-7, fibroblast growth factor 7HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGF-1, KGF-1/FGF-7, KGFfibroblast growth factor 7 (keratinocyte growth factor)
Gene Symbol FGF7
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C
Research Category Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein KGF/FGF-7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.