missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FGF7 Protein is fully biologically active when compared to standard. The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 10 ng/mL, corresponding to a specific activity of >1.0 x 10^5 IU/mg.
Specifications
Specifications
| Accession Number | P36363 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in 20 mM PB, pH 8.0, 1 M NaCl. |
| Gene ID (Entrez) | 2252 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 18.7 kDa |
| Quantity | 1000 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of KGF/FGF-7 as determined by LAL method. |
| Gene Alias | FGF-7, fibroblast growth factor 7HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGF-1, KGF-1/FGF-7, KGFfibroblast growth factor 7 (keratinocyte growth factor) |
| Gene Symbol | FGF7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?