missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Interleukin-23 subunit alpha Protein
A DNA sequence encoding the Mus musculus (Mouse) Interleukin-23 subunit alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8064-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_112542.1 | |
Interleukin-23 subunit alpha Protein | |
Research Use Only | |
Il23a | |
Recombinant Protein | |
E. coli | |
VPRSSSPDWA QCQQLSRNLC MLAWNAHAPA GHMNLLREEE DEETKNNVPR IQCEDGCDPQ GLKDNSQFCL QRIRQGLAFY KHLLDSDIFK GEPALLPDSP MEQLHTSLLG LSQLLQPEDH PRETQQMPSL SSSQQWQRPL LRSKILRSLQ AFLAIAARVF AHGAATLTEP LVPTA | |
Greater than 90% as determined by SDS-PAGE. |
83430 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |