missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Integrin alpha X beta 2 Protein
Description
The Recombinant Mouse Integrin alpha X beta 2 Protein is derived from CHO. The Recombinant Mouse Integrin alpha X beta 2 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Molecular Weight (g/mol) | 131 kDa (Integrin alphaX) & 83 kDa (Integrin beta2) |
| Name | Integrin alpha X beta 2 |
| Quantity | 50μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha X beta 2 protein, (N-terminus) Mouse Integrin alpha X (Phe20-Pro1116) Accession &Num; Q9QXH4 His-Pro GGGSGGGS Acidic Tail 6-His tag Mouse Integrin beta 2 (Gln24-Asn702) Accession &Num; P11835 His-Pro GGGSGGGS Basic Tail (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?