missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Integrin alpha 6 beta 1 Protein

Product Code. 18349287
Change view
Click to view available options
:
50ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18349287 50ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349287 Supplier R&D Systems™ Supplier No. 7810A6050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Integrin alpha 6 beta 1 Protein is derived from CHO. The Recombinant Mouse Integrin alpha 6 beta 1 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Molecular Weight (g/mol) 119 kDa ( alpha6 subunit) & 86.4 kDa (beta1 subunit)
Name Integrin alpha 6 beta 1
Quantity 50μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha 6 beta 1 protein, (N-terminus) Mouse Integrin alpha 6 (Phe24-Gly1011) Accession &Num; Q61739 HP GGGSGGGS Acidic Tail HHHHHH Mouse Integrin beta 1 (Gln21-Asp728) Accession &Num; P09055 HP GGGSGGGS Basic Tail (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Noncovalently-linked heterodimer
Gene Alias CD49f, ITGA6B, VLA-6
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.