missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Integrin alpha 4 beta 7 Protein

Product Code. 18370388
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370388 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370388 Supplier R&D Systems™ Supplier No. 8615A3050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Integrin alpha 4 beta 7 Protein is derived from CHO. The Recombinant Mouse Integrin alpha 4 beta 7 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 3676
Molecular Weight (g/mol) Observed M.W.: 115 - 170kDa, reducing conditions, Theoretical M.W.: 113kDa (Integrin alpha 4), 84kDa (Integrin beta 7)
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha 4 beta 7/LPAM-1 protein, (N-terminus) Mouse Integrin alpha 4 Tyr34-Gln970 (Arg591Leu) Accession &Num; AAH68313 GS Linker Acidic Tail HHHHHH Mouse Integrin ss7 Glu20-Arg724 Accession &Num; P26011 GS Linker Basic Tail (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49d/beta7, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), LPAM1, LPAM-1, MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Common Name Integrin alpha 4 beta 7/LPAM-1
Gene Symbol Itga5
Biological Activity Measured by the ability of the immobilized protein to support the adhesion of CHO Chinese hamster ovary cells transfected with VCAM-1. The ED50 for this effect is typically 0.6-3.6μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Mouse Integrin alpha 4 Tyr34-Gln970 (Arg591Leu) Accession # AAH68313, GS Linker, Acidic Tail, N-terminus; HHHHHH; Mouse Integrin ss7 Glu20-Arg724 Accession # P26011, GS Linker Basic Tail, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.