missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL7R / IL7RA Protein

Product Code. p-7152936
Click to view available options
Quantity:
100 μg
20 μg
50 μg
Unit Size:
100µg
20µg
50µg
This item is not returnable. View return policy

Product Code. 16081730

Brand: enQuireBio™ QP6226ba20ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) IL7R / IL7RA, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_032398.3
Gene ID (Entrez) 16197
Name IL7R / IL7RA Protein
Quantity 20 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Il7r
Protein Length Extracellular Domain
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Baculovirus
Protein Tag His
Sequence ESGNAQDGDL EDADADDHSF WCHSQLEVDG SQHLLTCAFN DSDINTANLE FQICGALLRV KCLTLNKLQD IYFIKTSEFL LIGSSNICVK LGQKNLTCKN MAINTIVKAE APSDLKVVYR KEANDFLVTF NAPHLKKKYL KKVKHDVAYR PARGESNWTH VSLFHTRTTI PQRKLRPKAM YEIKVRSIPH NDYFKGFWSE WSPSSTFETP EPKNQGGWD
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.