missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL1RL2 / IL-1Rrp2 Protein

Product Code. 16052851
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16052851

Brand: enQuireBio™ QP8137ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) IL1RL2 / IL-1Rrp2, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_573456.1
Gene ID (Entrez) 107527
Name IL1RL2 / IL-1Rrp2 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Il1rl2
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His
Sequence DTCEDIFMHN VIISEGQPFP FNCTYPPETN GAVNLTWYKT PSKSPVSNNR HLRVHQDQTW ILFLPLTLED SGIYQCVIRN AHNCYQIAVN LTVLKNHWCD SSMEGSPVNS PDVYQQILPI GKSGSLNCHL YFPESCALDS IKWYKGCEEI KAGKKYSPSG AKLLVNNVAV EDGGSYACSA RLTHLGRHFT IRNYIAVNTK EVEYGRRIPN ITYPKNNSIE VPLGSTLIVN CNITDTKENT NLRCWRVNNT LVDDYYKDSK RIQEGIETNV SLRDQIRYTV NITFLKVKME DYGRPFTCHA GVSAAYIILI YPVPDFR
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.