missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL17 / IL17A Protein

Product Code. 15986189
Click to view available options
Quantity:
1 mg
25 μg
5 μg
500 μg
Unit Size:
1mg
25µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 15986189

Brand: enQuireBio™ QP54665ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_034682.1
Gene ID (Entrez) 16171
Name IL17 / IL17A Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol IL17A
Biological Activity Shows bioactivity at a concentration of 5 ng/mL in the Production of IL-6 by mouse 3T3 cells assay. The bioactivity acceptance criteria is set at 2 x 10^5 units/mg.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence MAAIIPQSSA CPNTEAKDFL QNVKVNLKVF NSLGAKVSSR RPSDYLNRST SPWTLHRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPESCPF TFRVEKMLVG VGCTCVASIV RQAA
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.