missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse IL-7 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
IL7 Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine 2E8 cells is less than 0.2 ng/mL, corresponding to a specific activity of >5.0 x 10^6 IU/mg.
Specifications
Specifications
| Accession Number | Q544C8, Q544C8, Q544C8 |
| For Use With (Application) | Flow Cytometry, PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, 2 % trehalose. |
| Gene ID (Entrez) | 3574 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 14.9 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of IL-7 as determined by LAL method. |
| Gene Alias | IL-7interleukin-7, interleukin 7 |
| Gene Symbol | IL7 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction