missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-6R alpha (aa 20-357) Protein
Description
Measured by its binding ability in a functional ELISA. Immobilized Recombinant Mouse (rm) IL-6R alpha at 5μg/mL (100μL/well) on Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ) coated plates (0.25μg/well) can bind rmIL-6 with a linear range of 0.8-50 ng/mL.
The Recombinant Mouse IL-6R alpha (aa 20-357) Protein is derived from NS0. The Recombinant Mouse IL-6R alpha (aa 20-357) Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 16194 |
| Molecular Weight (g/mol) | 39 kDa (monomer) |
| Name | IL-6R alpha |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse IL-6R alpha protein Leu20-Glu357, with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Gene Alias | CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?