missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-6 protein is fully biologically active when compared to standard. The ED50 as determined by the dose-dependent stimulation of the proliferation of IL-6-dependent murine 7TD1 cells is less than 0.02 ng/mL, corresponding to a specific activity of >5 x 10^7 IU/mg
Specifications
Specifications
| Accession Number | P08505, P08505, P08505 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in 30 mM Acetic Acid, pH 3.0, 150 mM NaCl, 0.02 % Tween-20. |
| Gene ID (Entrez) | 3569 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 21.7 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of IL-6 as determined by LAL method. |
| Gene Alias | B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 |
| Gene Symbol | IL6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?