missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse IL-3R alpha/CD123 hIgG-His Protein

Product Code. 18351113 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
20 μg
250 μg
50 μg
Unit Size:
100µg
10µg
20µg
250µg
50µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351113 50 μg 50µg
18311715 250 μg 250µg
18314425 20 μg 20µg
18371587 10 μg 10µg
18330106 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 18351113 Supplier Novus Biologicals™ Supplier No. NBP31835950UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25 mg/ml
For Use With (Application) Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin, SDS-PAGE, Western Blot
Formulation 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 30% glycerol
Gene ID (Entrez) 3563
Molecular Weight (g/mol) M.W. Theoretical: 61.5 kDa
Quantity 50 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <1 EU per 1ug of protein (determined by LAL method)
Gene Alias CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Gene Symbol IL3RA
Research Category B Cell Development and Differentiation Markers, Cytokine Research, Oncogenes, Phospho Specific, Protein Kinase
Purity or Quality Grade >90%, by SDS-PAGE
Protein IL-3R alpha/CD123
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.