missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-36 alpha/IL-1F6 (aa 8-160) Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, TCEP and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, TCEP and EDTA with Trehalose.
Unit Size:
10µg
Description
The Recombinant Mouse IL-36 alpha/IL-1F6 (aa 8-160) Protein is derived from E. coli. The Recombinant Mouse IL-36 alpha/IL-1F6 (aa 8-160) Protein has been validated for the following applications: Bioactivity.
Tekniske data
Tekniske data
| Formulation | Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, TCEP and EDTA with Trehalose. |
| Gene ID (Entrez) | 54448 |
| Molecular Weight (g/mol) | 17 kDa |
| Name | IL-36 alpha/IL-1F6 (aa 8-160) |
| Quantity | 10 μg |
| Source | E. coli-derived mouse IL-36 alpha/IL-1F6 protein Arg8-His160 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion