missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-36 alpha/IL-1F6 (aa 8-160) Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, TCEP and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, TCEP and EDTA with Trehalose.
Unit Size:
10µg
Description
The Recombinant Mouse IL-36 alpha/IL-1F6 (aa 8-160) Protein is derived from E. coli. The Recombinant Mouse IL-36 alpha/IL-1F6 (aa 8-160) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in MOPS, NaCl, TCEP and EDTA with Trehalose. |
| Gene ID (Entrez) | 54448 |
| Molecular Weight (g/mol) | 17 kDa |
| Name | IL-36 alpha/IL-1F6 (aa 8-160) |
| Quantity | 10 μg |
| Source | E. coli-derived mouse IL-36 alpha/IL-1F6 protein Arg8-His160 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction