missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivning
IL3 Protein is fully biologically active when compared to standard. The ED50 as determined by the dose-dependent stimulation of the proliferation of murine M-NFS-60 cells is less than 0.05 ng/mL, corresponding to a specific activity of >2 x 10^7 IU/mg.
Specifikationer
Specifikationer
| Accession Number | P01586, P01586, P01586 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 3562 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 14.8 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of IL-3 as determined by LAL method. |
| Gene Alias | Hematopoietic growth factor, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor |
| Gene Symbol | IL3 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?