missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-28B/IFN-lambda 3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£327.21
Specifications
Gene ID (Entrez) | 338374 |
---|---|
Molecular Weight (g/mol) | 19.7 kDa |
Name | IL-28B/IFN-lambda 3 |
Quantity | 25μg |
Source | E. coli-derived mouse IL-28B/IFN-lambda 3 protein Asp20-Val193,with an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18177720
|
R&D Systems™
1789-ML-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£327.21
25µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18310328
|
R&D Systems™
1789-ML-025/CF |
£327.21
25µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Measured in an anti-viral assay using HepG2 human hepatocellular carcinoma cells infected with encephalomyocarditis (EMC) virus. Sheppard, P. et al. (2003) Nat. Immunol. 4:63. The ED50 for this effect is 0.800-8.00 ng/mL.Specifications
338374 | |
IL-28B/IFN-lambda 3 | |
E. coli-derived mouse IL-28B/IFN-lambda 3 protein Asp20-Val193,with an N-terminal Met | |
Recombinant |
19.7 kDa | |
25μg | |
Unconjugated |