missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-27 (NS0-expressed) Protein
Description
The Recombinant Mouse IL-27 (NS0-expressed) Protein is derived from NS0. The Recombinant Mouse IL-27 (NS0-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 246779 |
| Molecular Weight (g/mol) | 48.7 kDa |
| Name | IL-27 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse IL-27 protein, (N-terminus) Mouse EBI3 (Tyr19-Pro228) Accession &Num; O35228 GGGSGGGSGGGSGGGS Mouse p28 (Phe29-Ser234) Accession &Num; Q8K3I6 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?