missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-21 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£280.77
Specifications
Gene ID (Entrez) | 60505 |
---|---|
Molecular Weight (g/mol) | 14.4 kDa |
Name | IL-21 |
Quantity | 10μg |
Source | E. coli-derived mouse IL-21 protein Pro25-Ser146,with and without an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18177970
|
R&D Systems™
594-ML-010 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£280.77
10µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18117520
|
R&D Systems™
594-ML-010/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£280.77
10µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse IL-21 Protein is derived from E. coli. The Recombinant Mouse IL-21 Protein has been validated for the following applications: Bioactivity.Specifications
60505 | |
IL-21 | |
E. coli-derived mouse IL-21 protein Pro25-Ser146,with and without an N-terminal Met | |
Recombinant |
14.4 kDa | |
10μg | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |