missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-2 Protein
Click to view available options
Quantity:
100 μg
20 μg
500 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Sodium Citrate and Tween™-80 with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Sodium Citrate and Tween™-80.
Unit Size:
100µg
20µg
500µg
Description
The Recombinant Mouse IL-2 Protein is derived from E. coli. The Recombinant Mouse IL-2 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Sodium Citrate and Tween™-80 with BSA as a carrier protein. |
| Gene ID (Entrez) | 16183 |
| Molecular Weight (g/mol) | 17.2 kDa |
| Name | IL-2 |
| Quantity | 20 μg |
| Source | E. coli-derived mouse IL-2 protein Ala21-Gln169,with and without an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute at 100-200μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction