missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-1ra/IL-1F3/IL1RN protein is fully biologically active when compared to standard. The ED50 as determined by inhibiting IL-1alpha- dependent proliferation of murine D10S cells is less than 50 ng/mL, corresponding to a specific activity of >2.0 x 104 IU/mg in the presence of 50 pg/mL rHuIL-1alpha.
Specifications
Specifications
| Accession Number | Q542W1, Q542W1, Q542W1 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 3557 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 17.3 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of IL-1ra/IL-1F3/IL1RN as determined by LAL method. |
| Gene Alias | DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 |
| Gene Symbol | IL1RN |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?