missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-17D Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£321.81
Specifications
Gene ID (Entrez) | 239114 |
---|---|
Molecular Weight (g/mol) | 20 kDa (monomer) |
Name | IL-17D |
Quantity | 25μg |
Source | E. coli-derived mouse IL-17D protein Ala25-Arg205 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18127350
|
R&D Systems™
2274-ML-025 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
£321.81
25µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18136690
|
R&D Systems™
2274-ML-025/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
£321.81
25µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse IL-17D Protein is derived from E. coli. The Recombinant Mouse IL-17D Protein has been validated for the following applications: Binding Activity.Specifications
239114 | |
IL-17D | |
E. coli-derived mouse IL-17D protein Ala25-Arg205 | |
Disulfide-linked homodimer | |
Recombinant |
20 kDa (monomer) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |