missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-17B Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£327.21
Specifications
Gene ID (Entrez) | 56069 |
---|---|
Molecular Weight (g/mol) | 18.2 kDa (monomer) |
Name | IL-17B |
Quantity | 25μg |
Source | E. coli-derived mouse IL-17B protein His21-Phe180,with an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18117380
|
R&D Systems™
1709-ML-025 |
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 and NaCl with BSA as a carrier protein. |
£327.21
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18177330
|
R&D Systems™
1709-ML-025/CF |
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 and NaCl. |
£327.21
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse IL-17B Protein is derived from E. coli. The Recombinant Mouse IL-17B Protein has been validated for the following applications: Bioactivity.Specifications
56069 | |
IL-17B | |
E. coli-derived mouse IL-17B protein His21-Phe180,with an N-terminal Met | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
18.2 kDa (monomer) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |