missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-17A/F Heterodimer Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Unit Size:
25µg
Description
The Recombinant Mouse IL-17A/F Heterodimer Protein is derived from E. coli. The Recombinant Mouse IL-17A/F Heterodimer Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
| Molecular Weight (g/mol) | 14.9 kDa (IL-17A) & 15.5 kDa (IL-17F), reducing conditions |
| Name | IL-17A/F Heterodimer |
| Quantity | 25 μg |
| Source | E. coli-derived mouse IL-17A/F Heterodimer protein, (N-terminus) Mouse IL-17A (Thr22-Ala158) Accession &Num; Q62386 Mouse IL-17F (Arg21-Ala153,with an N-terminal Met) Accession &Num; AAM77568 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction