missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-13 R alpha 1 Fc Chimera Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
200µg
Description
The Recombinant Mouse IL-13 R alpha 1 Fc Chimera Protein is derived from NS0. The Recombinant Mouse IL-13 R alpha 1 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 16164 |
| Molecular Weight (g/mol) | 63 kDa (monomer) |
| Name | IL-13R alpha 1 |
| Quantity | 200 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse IL-13 R alpha 1 protein, (N-terminus) Mouse IL-13 R alpha 1 (Ala25-Thr340) Accession &Num; O09030 DIEGR Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction