missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-13 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£258.09 - £749.44
Specifications
Gene ID (Entrez) | 16163 |
---|---|
Molecular Weight (g/mol) | 11.5 kDa |
Name | IL-13 |
Source | E. coli-derived mouse IL-13 protein Ser26-Phe131 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18147390
|
R&D Systems™
413-ML-005 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 5μg |
£258.09
5µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18186450
|
R&D Systems™
413-ML-005/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 5μg |
£258.09
5µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18176860
|
R&D Systems™
413-ML-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 25μg |
£749.44
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18398937
|
R&D Systems™
413-ML-025/CF |
£749.44
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse IL-13 Protein is derived from E. coli. The Recombinant Mouse IL-13 Protein has been validated for the following applications: Bioactivity.Specifications
16163 | |
IL-13 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
11.5 kDa | |
E. coli-derived mouse IL-13 protein Ser26-Phe131 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |