missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-12 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ 419-ML-050

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18359677

  • £667.37 / 50µg
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse IL-12 Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse IL-12 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
IL-12
Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse IL-12 protein, (N-terminus) Mouse IL-12 p40 (Met23-Ser335) Accession &Num; P43432 Mouse IL-12 p35 (Arg23-Ala215) Accession &Num; NP_032377 (C-terminus)
Disulfide-linked heterodimer
Recombinant
16159
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers