missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-12 (linked heterodimer) Protein

Product Code. 18756496
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18756496

Brand: R&D Systems™ 10051ML010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Recombinant Mouse IL-12 (linked heterodimer) Protein is derived from NS0

Measured in a cell proliferation assay using PHA-activated mouse splenocytes. Mattner, F. et al. (1993) Eur. J. Immunol. 23:2202. The ED50 for this effect is 0.01-0.1 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43432, P43431
For Use With (Application) Bioactivity
Gene ID (Entrez) 3592
Molecular Weight (g/mol) 58 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 10 μg
Source

Mouse myeloma cell line, NS0-derived mouse IL-12 protein

Human IL-12 p40 (Met23-Ser335) Accession # P43432GGGGSGGGGSGGGGSMouse IL-12 p35 (Arg23-Ala215) Accession # P43431N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.