missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-11 Protein

Product Code. 18380298
Click to view available options
Quantity:
25 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
PBS, pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl), 0.1% BSA
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18380298

Brand: R&D Systems™ 418ML025

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse IL-11 Protein is derived from E. coli. The Recombinant Mouse IL-11 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation PBS, pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl), 0.1% BSA
Gene ID (Entrez) 16156
Molecular Weight (g/mol) 19 kDa
Name IL-11
Quantity 25 μg
Source E. coli-derived mouse IL-11 protein Gly23-Leu199,with an N-terminal Met and Pro25-Leu199
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias Adipogenesis inhibitory factor, AGIF, AGIFoprelvekin, IL11, IL-11Oprelvekin, interleukin 11, interleukin-11, Oprelvekin
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile, deionized water.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.