missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-1 beta/IL-1F2 Protein

Product Code. 18187330
Change view
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in PBS.
Unit Size:
25µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Formulation unitSize
18187330 5 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5µg
18354947 5 μg Supplied as a 0.2μm filtered solution in PBS. 5µg
18118290 25 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25µg
18319407 25 μg Supplied as a 0.2μm filtered solution in PBS. 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18187330 Supplier R&D Systems™ Supplier No. 401ML005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse IL-1 beta/IL-1F2 Protein is derived from E. coli. The Recombinant Mouse IL-1 beta/IL-1F2 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 16176
Molecular Weight (g/mol) 17 kDa
Name IL-1 beta/IL-1F2
Quantity 5 μg
Source E. coli-derived mouse IL-1 beta/IL-1F2 protein Val118-Ser269,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.