missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IL-1 alpha/IL-1F1 Protein

Product Code. 18178250
Change view
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18178250 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18178250 Supplier R&D Systems™ Supplier No. 400ML005/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse IL-1 alpha/IL-1F1 Protein is derived from E. coli. The Recombinant Mouse IL-1 alpha/IL-1F1 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 16175
Molecular Weight (g/mol) 18 kDa
Name IL-1 alpha/IL-1F1
Quantity 5 μg
Source E. coli-derived mouse IL-1 alpha/IL-1F1 protein Ser6-Ser161
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 50μg/mL in sterile PBS.
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.