missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recombinant Mouse IgG2A Fc Protein
Description
Bioassay data are not available.
Specifications
Specifications
| Accession Number | P01863 |
| Concentration | LYOPH |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Molecular Weight (g/mol) | M.W. (Observed): 34-35 kDa, reducing conditions; M.W. (theoretical): 26.7 kDa (monomer) |
| Quantity | 100 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse IgG2A protein Glu98-Lys330, with an N-terminal Met-Asp-Pro |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.01 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?