missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IGFBP-3 Protein
Description
The Recombinant Mouse IGFBP-3 Protein is derived from NS0. The Recombinant Mouse IGFBP-3 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | CAA57271.1 |
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
| Gene ID (Entrez) | 16009 |
| Molecular Weight (g/mol) | 29.4 kDa |
| Name | IGFBP-3 |
| Quantity | 25μg |
| Source | Mouse myeloma cell line,NS0-derived mouse IGFBP-3 protein Pro22-Gln291 with substitutions Arg250Gln,Gln259Arg,Ser260Gly,Arg271Pro |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?