missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IGF1 / IGF-I Protein
Recombinant Protein
Brand: enQuireBio™ QP5404-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001104744.1 | |
IGF1 / IGF-I Protein | |
Research Use Only | |
IGF1 | |
Recombinant Protein | |
E. coli | |
GPETLCGAEL VDALQFVCGP RGFYFNKPTG YGSSIRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPTKAA | |
Greater than 95% as determined by SDS-PAGE. |
16000 | |
50 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 20 ng/mL in the FDC-P1 cell proliferation assay. The bioactivity acceptance criteria is set at 5.0 x 10^4 units/mg. | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |