missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IGF1 / IGF-I Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
50µg
Specifications
Specifications
| Accession Number | NP_001104744.1 |
| Gene ID (Entrez) | 16000 |
| Name | IGF1 / IGF-I Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | IGF1 |
| Biological Activity | Shows bioactivity at a concentration of 20 ng/mL in the FDC-P1 cell proliferation assay. The bioactivity acceptance criteria is set at 5.0 x 10^4 units/mg. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Mouse |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction