missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IFNG / Interferon Gamma Protein

Product Code. 15975579
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15975579 20 μg 20µg
15955579 100 μg 100µg
15985579 250 μg 250µg
15995579 500 μg 500µg
15965579 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15975579 Supplier enQuireBio™ Supplier No. QP414120ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_032363.1
Gene ID (Entrez) 15978
Name IFNG / Interferon Gamma Protein
Quantity 20 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol IFNG
Biological Activity Shows bioactivity in the Viral CPE assay using EMC virus on L929 cells assay. The bioactivity acceptance criteria is set at 1 x 10^7 units/mg.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence MHGTVIESLE SLNNYFNSSG IDVEEKSLFL DIWRNWQKDG DMKILQSQII SFYLRLFEVL KDNQAISNNI SVIESHLITT FFSNSKAKKD AFMSIAKFEV NNPQVQRQAF NELIRVVHQL LPESSLRKRK RSRC
Buffer 10 mM sodium phosphate, 100 mM sodium chloride, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.