missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse IFN-alpha 13 Protein

Product Code. 18789456
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
This item is not returnable. View return policy

Product Code. 18789456

Brand: R&D Systems™ 10481IF010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured in an anti-viral assay using L-929 mouse fibroblast cells infected with encephalomyocarditis (EMC) virus. Vogel, S.N. et al. (1982) Infect. Immunol. 38:681. The ED50 for this effect is 1-15 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q80SU4.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3447
Molecular Weight (g/mol) M.W.-observed: 20-30 kDa, under reducing conditions, M.W.-theoretical: 19 kDa
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived mouse IFN-alpha 13 protein Gly23-Glu189
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias IFL, IFNA1, IFNA13, IFNalpha 13, IFN-alpha 1 b, IFN-alpha-1/13, interferon alpha-1/13, Interferon alpha-D, interferon, alpha 13, LeIF D
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IFN-alpha 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.