missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse ICAM-2/CD102 Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£233.26 - £641.45
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in Tris, Citric Acid and NaCl. |
---|---|
Gene ID (Entrez) | 15896 |
Molecular Weight (g/mol) | 49.5 kDa (monomer) |
Name | ICAM-2/CD102 Fc Chimera |
Source | Mouse myeloma cell line,NS0-derived mouse ICAM-2/CD102 protein, (N-terminus) Mouse ICAM-2 (Ser20-Gln222) Accession &Num; P35330 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18147740
|
R&D Systems™
774-M2-200 |
200μg |
£641.45
200µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18197320
|
R&D Systems™
774-M2-050 |
50μg |
£233.26
50µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse ICAM-2/CD102 Fc Chimera Protein is derived from NS0. The Recombinant Mouse ICAM-2/CD102 Fc Chimera Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Tris, Citric Acid and NaCl. | |
49.5 kDa (monomer) | |
Mouse myeloma cell line,NS0-derived mouse ICAM-2/CD102 protein, (N-terminus) Mouse ICAM-2 (Ser20-Gln222) Accession &Num; P35330 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
15896 | |
ICAM-2/CD102 Fc Chimera | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |