missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse ICAM-1/CD54 Fc Chimera Protein

Product Code. 18378957
Change view
Click to view available options
:
50ug; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18378957 50ug; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378957 Supplier R&D Systems™ Supplier No. 796IC050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by the ability of the immobilized protein to support the adhesion of PMA-stimulated HSB2 human peripheral blood acute lymphoblastic leukemia cells. When 5 x 104 cells/well are added to rmICAM/Fc Chimera coated plates (12.5 μg/mL with 100 μL/well), approximately 65-95% will adhere after 1 hour incubation at 37 °C. Optimal dilutions should be determined by each laboratory for each application.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q3U8M7
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3383
Molecular Weight (g/mol) M.W. (Observed): 110-130 kDa, reducing conditions; M.W. (theoretical): 76.8 kDa (monomer)
Name ICAM-1/CD54 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse ICAM-1/CD54 protein Mouse ICAM-1 (Gln28-Asn485) (N-terminus) Accession # Q3U8M7 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Icam1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 0.4 mg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ICAM-1/CD54
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.