missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse HVEM/TNFRSF14 Fc Chimera Protein

Product Code. 18337427
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337427 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337427 Supplier R&D Systems™ Supplier No. 2516HV100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. When Recombinant Mouse (rm) HVEM Fc Chimera is immobilized at 100 ng/ mL, 100 μL/well, the concentration of biotinylated rmBTLA Fc Chimera that produces 50% of the optimal binding response is found to be approximately 30-120 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_849262
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 8764
Molecular Weight (g/mol) M.W. (Observed): 60-65 kDa, reducing conditions; M.W. (theoretical): 45.6 kDa
Name HVEM/TNFRSF14 Fc Chimera
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse HVEM/TNFRSF14 protein Mouse HVEM (Gln39-Val207) (N-terminus) Accession # NP_849262 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol Tnfrsf14
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 1 mg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein HVEM/TNFRSF14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.