missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse HGF Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity
£358.52
Specifications
Gene ID (Entrez) | 15234 |
---|---|
Molecular Weight (g/mol) | 53.4 kDa ( alpha-chain), 26 kDa (beta-chain) |
Name | HGF |
Quantity | 25μg |
Source | Mouse myeloma cell line,NS0-derived mouse HGF protein Gln33-Arg495 (alpha) & Val496-Leu728 (beta) |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18117410
|
R&D Systems™
2207-HG-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£358.52
25µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18338967
|
R&D Systems™
2207-HG-025/CF |
£358.52
25µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse HGF Protein is derived from NS0. The Recombinant Mouse HGF Protein has been validated for the following applications: Bioactivity, Binding Activity.Specifications
15234 | |
HGF | |
Mouse myeloma cell line,NS0-derived mouse HGF protein Gln33-Arg495 (alpha) & Val496-Leu728 (beta) | |
Disulfide-linked heterodimer | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
53.4 kDa ( alpha-chain), 26 kDa (beta-chain) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A | |
Recombinant |