missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse HB-EGF Protein

Product Code. 18400872 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400872 500 μg 500µg
18436251 10 μg 10µg
18443202 100 μg 100µg
18421772 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18400872 Supplier Novus Biologicals™ Supplier No. NBP235069500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

HB-EGF protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 1 ng/mL, corresponding to a specific activity of >1.0 x 106IU/mg.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q06186, Q06186, Q06186
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 10 mM PB, 500 mM NaCl, pH7.4.
Gene ID (Entrez) 1839
Molecular Weight (g/mol) 9.8 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of HB-EGF as determined by LAL method.
Gene Alias diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), DTS, DTSF, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor
Gene Symbol HBEGF
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Biologically Active Proteins
Purity or Quality Grade >97% pure by SDS-PAGE and HPLC
Protein HB-EGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.