missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse HABP1/C1QBP Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Mouse HABP1/C1QBP Protein is derived from E. coli. The Recombinant Mouse HABP1/C1QBP Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in MOPS and NaCl with Trehalose. |
| Gene ID (Entrez) | 12261 |
| Molecular Weight (g/mol) | 24.8 kDa |
| Name | HABP1/C1QBP |
| Quantity | 50 μg |
| Source | E. coli-derived mouse HABP1/C1QBP protein, (N-terminus) Met-Met Mouse HABP1 (Leu72-Gln279) Accession &Num; NP_031599 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur