missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse H2-D1 Protein
A DNA sequence encoding the Mus musculus (Mouse) H2-D1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7403-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
H2-D1 Protein | |
Research Use Only | |
H2-D1 | |
Recombinant Protein | |
E. coli | |
GSHSLRYFVT AVSRPGFGEP RYMEVGYVDN TEFVRFDSDA ENPRYEPRAR WIEQEGPEYW ERETRRAKGN EQSFRVDLRT ALRYYNQSAG GSHTLQWMAG CDVESDGRLL RGYWQFAYDG CDYIALNEDL KTWTAADMAA QITRRKWEQA GAAERDRAYL EGECVEWLRR YLKNGNATLL RTDPPKAHVT HHRRPEGDVT LRCWALGFYP ADITLTWQLN GEELTQEMEL VETRPAGDGT FQKWASVVVP LGKEQKYTCH VEHEGLPEPL TLRWGKEEPP SSTKTNT | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |