missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GSTP2 Protein

A cDNA sequence encoding the GSTP2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12063-HIS-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939248

  • £99.15 / 2µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

14869
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMPPYTIV YFPSPGRCEA MRMLLADQGQ SWKEEVVTID TWMQGLLKPT CLYGQLPKFE DGDLTLYQSN AILRHLGRSL GLYGKNQREA AQVDMVNDGV EDLRGKYGTM IYRNYENGKN DYVKALPGHL KPFETLLSQN QGGKAFIVGD QISFADYNLL DLLLIHQVLA PGCLDNFPLL SAYVARLSAR PKIKAFLSSP EHVNRPINGN GKQ
Greater than 95.0% as determined by SDS-PAGE.
GSTP2 Protein
Research Use Only
Gstp2
Mouse
His
GSTP2 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers