missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse GSTA1 His Protein

Product Code. 18295020 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.1mg
0.50mg
This item is not returnable. View return policy

Product Code. 18295020

Brand: Novus Biologicals™ NBP2520890.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-223 of Mouse GSTA1 The Recombinant Mouse GSTA1 Protein is derived from E. coli. The Recombinant Mouse GSTA1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH7.4) containing 10% glycerol, 1mM DTT
Gene ID (Entrez) 2938
Molecular Weight (g/mol) TMW: 28kDa
Quantity 0.5 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias EC 2.5.1.18, glutathione S-alkyltransferase A1, glutathione S-aryltransferase A1, glutathione S-transferase 2, glutathione S-transferase A1, glutathione S-transferase alpha 1, glutathione S-transferase Ha subunit 1, GST class-alpha member 1, GST HA subunit 1, GST, class alpha, 1, GST2, GSTA1-1, GST-epsilon, GTH1, MGC131939, S-(hydroxyalkyl)glutathione lyase A1
Common Name GSTA1
Biological Activity Specific activity is > 4,000 pmol/min/ug, and is defined as the amount of enzyme that conjugate 1.0 pmole of 1-chloro-2,4-dinitrobenzene (CDNB) with reduced glutathione per minute at pH 6.5 at 25C.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.