missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Granzyme D Protein
Description
The Recombinant Mouse Granzyme D Protein is derived from NS0. The Recombinant Mouse Granzyme D Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in HEPES and NaCl. |
| Gene ID (Entrez) | 14941 |
| Molecular Weight (g/mol) | 27 kDa |
| Name | Granzyme D |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Granzyme D protein Ile21-Leu252 with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | CCP5, Ctla5, Ctla-5, granzyme D, GrzD |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?